1-Chloro-2,4-difluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

51258-1G 51258-25G 51258-500MG 51258-5G
76891-014EA 129.43 USD
76891-014 76891-016 76891-018 76891-020
1-Chloro-2,4-difluoro-3-iodobenzene ≥97%
1-Chloro-2,4-difluoro-3-iodobenzene
Formula: C₆H₂ClF₂I
Storage Temperature: Ambient
MDL Number: MFCD12922594
CAS Number: 1208078-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR