1-Bromo-4-((4-chlorophenoxy)methyl)benzene ≥97%

Supplier: AOBChem USA

Warning

49581-1G 49581-25G 49581-500MG 49581-5G
76890-786EA 233.4 USD
76890-786 76890-788 76890-790 76890-792
1-Bromo-4-((4-chlorophenoxy)methyl)benzene ≥97%
1-Bromo-4-((4-chlorophenoxy)methyl)benzene
Formula: C₁₃H₁₀BrClO
MW: 297.575 g/mol
Storage Temperature: Ambient
MDL Number: MFCD3880423
CAS Number: 884162-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR