2-(3-Bromophenyl)benzothiazole ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-(3-Bromophenyl)benzo[d]thiazole

36296-1G 36296-25G 36296-500MG 36296-5G
76888-300EA 236.59 USD
76888-300 76888-302 76888-304 76888-306
2-(3-Bromophenyl)benzothiazole ≥97% research grade
2-(3-Bromophenyl)benzothiazole
Formula: C₁₃H₈BrNS
Storage Temperature: Ambient
MDL Number: MFCD00579076
CAS Number: 19654-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR