1-Bromo-2,3-dichloro-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

34648-25G 34648-5G
76887-886EA 481.65 USD
76887-886 76887-888
1-Bromo-2,3-dichloro-4-fluorobenzene ≥97%
1-Bromo-2,3-dichloro-4-fluorobenzene
Formula: C₆H₂BrCl₂F
Storage Temperature: Ambient
MDL Number: MFCD11845895
CAS Number: 1093092-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR