1-Benzyloxy-2-bromo-6-chloro-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

33548-25G 33548-5G
76887-616EA 453.01 USD
76887-616 76887-618
1-Benzyloxy-2-bromo-6-chloro-4-fluorobenzene ≥97%
1-Benzyloxy-2-bromo-6-chloro-4-fluorobenzene
Formula: C₁₃H₉BrClFO
MW: 315.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834078
CAS Number: 2121512-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR