1-(5-Bromo-2-fluoro-4-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

26407-1G 26407-25G 26407-500MG 26407-5G
76885-986EA 266.29 USD
76885-986 76885-988 76885-990 76885-992
1-(5-Bromo-2-fluoro-4-methoxyphenyl)ethanone ≥97%
1-(5-Bromo-2-fluoro-4-methoxyphenyl)ethanone
Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Ambient
CAS Number: 914221-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR