2-(3,5-Dichlorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

19643-25G 19643-5G
76884-330EA 537.88 USD
76884-330 76884-332
2-(3,5-Dichlorophenyl)-1,3-dioxolane ≥97%
2-(3,5-Dichlorophenyl)-1,3-dioxolane
Formula: C₉H₈Cl₂O₂
MW: 219.065 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06208944
CAS Number: 70185-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR