5-Chloro-4-fluoro-2-hydroxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

33860-1G 33860-25G 33860-500MG 33860-5G
76931-306EA 380.87 USD
76931-306 76931-308 76931-310 76931-312
5-Chloro-4-fluoro-2-hydroxybenzaldehyde ≥95%
5-Chloro-4-fluoro-2-hydroxybenzaldehyde
Formula: C₇H₄ClFO₂
MW: 174.557 g/mol
Storage Temperature: Ambient
CAS Number: 264879-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR