2-Bromo-4-(tert-butyl)-1-chlorobenzene ≥95%

Supplier: AOBChem USA

Warning

48439-1G 48439-25G 48439-5G
76904-316EA 66.84 USD
76904-316 76904-318 76904-320
2-Bromo-4-(tert-butyl)-1-chlorobenzene ≥95%
2-Bromo-4-(tert-butyl)-1-chlorobenzene
Formula: C₁₀H₁₂BrCl
Storage Temperature: Refrigerator
MDL Number: MFCD09878145
CAS Number: 61024-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR