2-Bromo-6-ethylpyridine ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-6-ethyl-pyridine

21266-1G 21266-25G 21266-500MG 21266-5G
76898-350EA 304.48 USD
76898-350 76898-352 76898-354 76898-356
2-Bromo-6-ethylpyridine ≥95% research grade
2-Bromo-6-ethylpyridine
Formula: C₇H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD11869660
CAS Number: 83004-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR