2-Benzyloxy-1-bromo-3-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

16732-1G 16732-25G 16732-5G
76896-546EA 82.76 USD
76896-546 76896-548 76896-550
2-Benzyloxy-1-bromo-3-fluorobenzene ≥95%
2-Benzyloxy-1-bromo-3-fluorobenzene
Formula: C₁₃H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD11520674
CAS Number: 1036724-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR