4-Bromo-1-isopropoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

23747-1G 23747-25G 23747-5G
76911-356EA 67.9 USD
76911-356 76911-358 76911-360
4-Bromo-1-isopropoxy-2-methylbenzene ≥95%
4-Bromo-1-isopropoxy-2-methylbenzene
Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547897
CAS Number: 749927-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR