1-(4-Bromo-2-fluorophenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

18160-1G 18160-25G 18160-500MG 18160-5G
76883-926EA 218.55 USD
76883-926 76883-928 76883-930 76883-932
1-(4-Bromo-2-fluorophenyl)ethanol ≥95%
1-(4-Bromo-2-fluorophenyl)ethanol
Formula: C₈H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD19601833
CAS Number: 353282-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR