Methyl 6-bromo-4-fluorobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

99064-1G 99064-25G 99064-500MG 99064-5G
76944-160EA 165.51 USD
76944-160 76944-162 76944-164 76944-166
Methyl 6-bromo-4-fluorobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 6-bromo-4-fluorobenzo[b]thiophene-2-carboxylate
Formula: C₁₀H₆BrFO₂S
MW: 289.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044572
CAS Number: 154650-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR