N-(4-Bromo-2,5-dimethylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

90725-25G 90725-5G
76944-126EA 488.02 USD
76944-126 76944-128
N-(4-Bromo-2,5-dimethylphenyl)acetamide ≥97%
N-(4-Bromo-2,5-dimethylphenyl)acetamide
Formula: C₁₀H₁₂BrNO
MW: 242.112 g/mol
Storage Temperature: Ambient
CAS Number: 13711-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR