9-Ethyl-9H-carbazole-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

88409-1G 88409-25G 88409-5G
76944-116EA 103.97 USD
76944-116 76944-118 76944-120
9-Ethyl-9H-carbazole-3-carboxylic acid ≥97%
9-Ethyl-9H-carbazole-3-carboxylic acid
Formula: C₁₅H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD06203133
CAS Number: 57102-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR