Trimethyl(4-propylphenyl)silane ≥97%

Supplier: AOBChem USA

Warning

86146-1G 86146-25G 86146-500MG 86146-5G
76944-086EA 318.27 USD
76944-086 76944-088 76944-090 76944-092
Trimethyl(4-propylphenyl)silane ≥97%
Trimethyl(4-propylphenyl)silane
Formula: C₁₂H₂₀Si
Storage Temperature: Ambient
CAS Number: 81631-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR