6,7-Dibromobenzo(1,4)dioxan ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 6,7-Dibromo-2,3-dihydrobenzo[b][1,4]dioxine

83857-25G 83857-5G
76944-074EA 418 USD
76944-074 76944-076
6,7-Dibromobenzo(1,4)dioxan ≥97% research grade
6,7-Dibromobenzo(1,4)dioxan
Formula: C₈H₆Br₂O₂
MW: 293.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463506
CAS Number: 25812-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR