Ethyl 2-(3-bromo-5-methylphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82659-1G 82659-25G 82659-500MG 82659-5G
76944-042EA 292.81 USD
76944-042 76944-044 76944-046 76944-048
Ethyl 2-(3-bromo-5-methylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(3-bromo-5-methylphenyl)-2-oxoacetate
Formula: C₁₁H₁₁BrO₃
MW: 271.107 g/mol
Storage Temperature: Ambient
CAS Number: 1892111-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR