Ethyl 2-(4-chloro-3-ethylphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82513-1G 82513-25G 82513-500MG 82513-5G
76944-026EA 341.61 USD
76944-026 76944-028 76944-030 76944-032
Ethyl 2-(4-chloro-3-ethylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(4-chloro-3-ethylphenyl)-2-oxoacetate
Formula: C₁₂H₁₃ClO₃
MW: 240.683 g/mol
Storage Temperature: Ambient
CAS Number: 1373519-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR