Ethyl 2-(2,5-dimethylphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82438-25G 82438-5G
76944-014EA 318.27 USD
76944-014 76944-016
Ethyl 2-(2,5-dimethylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(2,5-dimethylphenyl)-2-oxoacetate
Formula: C₁₂H₁₄O₃
MW: 206.238 g/mol
Storage Temperature: Ambient
CAS Number: 80120-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR