Ethyl-4-(dimethylamino)benzoylformate ≥97%

Supplier: AOBChem USA

Warning

82435-25G 82435-5G
76944-004EA 378.75 USD
76944-004 76944-006
Ethyl-4-(dimethylamino)benzoylformate ≥97%
Ethyl 4-(dimethylamino)benzoylformate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
Storage Temperature: Ambient
CAS Number: 41116-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR