Ethyl-5-fluoro-2-methoxybenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 5-fluoro-2-methoxybenzoylformate, Ethyl 2-(5-fluoro-2-methoxyphenyl)-2-oxoacetate

82410-1G 82410-25G 82410-5G
76943-976EA 109.28 USD
76943-976 76943-978 76943-980
Ethyl-5-fluoro-2-methoxybenzoylformate ≥95%
Ethyl-5-fluoro-2-methoxybenzoylformate
Formula: C₁₁H₁₁FO₄
MW: 226.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801422
CAS Number: 873548-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR