Ethyl-3-fluorobenzoylformate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-fluorophenyl)-2-oxoacetate

82402-1G 82402-25G 82402-500MG 82402-5G
76943-964EA 192.03 USD
76943-964 76943-966 76943-968 76943-970
Ethyl-3-fluorobenzoylformate ≥95%
Ethyl-3-fluorobenzoylformate
Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319618
CAS Number: 110193-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR