Ethyl-2-fluorobenzoylformate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(2-fluorophenyl)-2-oxoacetate

82393-1G 82393-25G 82393-5G
76943-938EA 93.36 USD
76943-938 76943-940 76943-942
Ethyl-2-fluorobenzoylformate ≥97%
Ethyl 2-fluorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD09801420
CAS Number: 1813-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR