tert-Butyl 2,5-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

80458-1G 80458-25G 80458-5G
76943-924EA 79.57 USD
76943-924 76943-926 76943-928
tert-Butyl 2,5-dichlorobenzoate ≥97%
tert-Butyl 2,5-dichlorobenzoate
Formula: C₁₁H₁₂Cl₂O₂
MW: 247.118 g/mol
Storage Temperature: Refrigerator
CAS Number: 127715-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR