tert-Butyl (3,4-difluorophenyl)carbamate ≥97% research grade

Supplier: AOBChem USA

Warning

80356-1G 80356-25G 80356-5G
76943-912EA 64.72 USD
76943-912 76943-914 76943-916
tert-Butyl (3,4-difluorophenyl)carbamate ≥97% research grade
tert-Butyl (3,4-difluorophenyl)carbamate
Formula: C₁₁H₁₃F₂NO₂
Storage Temperature: Ambient
CAS Number: 144298-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR