N-Boc-2-fluoroaniline ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: tert-Butyl (2-fluorophenyl)carbamate

80247-25G 80247-5G
76943-900EA 598.35 USD
76943-900 76943-902
N-Boc-2-fluoroaniline ≥95% research grade
N-Boc-2-fluoroaniline
Formula: C₁₁H₁₄FNO₂
MW: 211.2300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112559
CAS Number: 98968-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR