Methyl 2-cyano-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

76895-25G 76895-5G
76943-876EA 474.23 USD
76943-876 76943-878
Methyl 2-cyano-5-methoxybenzoate ≥97%
Methyl 2-cyano-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.183 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09800435
CAS Number: 127510-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR