N-(3-Fluoro-2-methylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

73103-1G 73103-25G 73103-5G
76943-840EA 102.91 USD
76943-840 76943-842 76943-844
N-(3-Fluoro-2-methylphenyl)acetamide ≥97%
N-(3-Fluoro-2-methylphenyl)acetamide
Formula: C₉H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD03787362
CAS Number: 322-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR