p-Tolyltrimethylsilane ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Trimethyl-p-tolylsilane, Trimethyl(p-tolyl)silane

68832-25G 68832-5G
76943-806EA 289.63 USD
76943-806 76943-808
p-Tolyltrimethylsilane ≥95%
p-Tolyltrimethylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₆Si
MW: 164.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014850
CAS Number: 3728-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR