Ethyl 2,6-dichloro-4-methylnicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

66028-1G 66028-25G 66028-5G
76943-796EA 88.06 USD
76943-796 76943-798 76943-800
Ethyl 2,6-dichloro-4-methylnicotinate ≥97% research grade
Ethyl 2,6-dichloro-4-methylnicotinate
Formula: C₉H₉Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD21604262
CAS Number: 108130-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR