tert-Butyl (4-bromo-2-fluoro-5-methylphenyl)carbamate ≥97%

Supplier: AOBChem USA

Warning

65140-1G 65140-25G 65140-500MG 65140-5G
76943-772EA 164.44 USD
76943-772 76943-774 76943-776 76943-778
tert-Butyl (4-bromo-2-fluoro-5-methylphenyl)carbamate ≥97%
tert-Butyl (4-bromo-2-fluoro-5-methylphenyl)carbamate
Formula: C₁₂H₁₅BrFNO₂
MW: 304.1554 g/mol
Storage Temperature: Ambient
CAS Number: 1260804-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR