tert-Butyl-6-bromopyridin-2-ylcarbamate ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
tert-Butyl (6-bromopyridin-2-yl)carbamate, N-Boc-2-Amino-6-bromopyridine
64933-25G
64933-5G
76943-762EA
481.65
USD
76943-762
76943-764
tert-Butyl-6-bromopyridin-2-ylcarbamate ≥97% research grade
tert-Butyl-6-bromopyridin-2-ylcarbamate
Formula:
C₁₀H₁₃BrN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD07367929 CAS Number: 344331-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...