Ethyl 3-bromo-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

62308-1G 62308-25G 62308-5G
76943-674EA 88.06 USD
76943-674 76943-676 76943-678
Ethyl 3-bromo-5-methylbenzoate ≥97%
Ethyl 3-bromo-5-methylbenzoate
Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
CAS Number: 612834-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR