Methyl 3-bromo-2-fluoro-6-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

61624-1G 61624-500MG 61624-5G
76943-644EA 693.83 USD
76943-644 76943-646 76943-648
Methyl 3-bromo-2-fluoro-6-methylbenzoate ≥97%
Methyl 3-bromo-2-fluoro-6-methylbenzoate
Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Ambient
CAS Number: 1513482-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR