6-Chloro-N-methyl-2-pyridinamine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-N-methylpyridin-2-amine

61165-25G 61165-5G
76943-618EA 389.36 USD
76943-618 76943-620
6-Chloro-N-methyl-2-pyridinamine ≥97%
6-Chloro-N-methyl-2-pyridinamine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234196
CAS Number: 89026-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR