6-Bromo-2,3,4-trifluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

61076-1G 61076-25G 61076-500MG 61076-5G
76943-602EA 134.74 USD
76943-602 76943-604 76943-606 76943-608
6-Bromo-2,3,4-trifluorobenzoic acid ≥97% research grade
6-Bromo-2,3,4-trifluorobenzoic acid
Formula: C₇H₂BrF₃O₂
MW: 254.989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09910412
CAS Number: 530145-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR