Methyl 3-formyl-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

60792-1G 60792-25G 60792-500MG 60792-5G
76943-592EA 227.04 USD
76943-592 76943-594 76943-596 76943-598
Methyl 3-formyl-2-methoxybenzoate ≥97%
Methyl 3-formyl-2-methoxybenzoate
Formula: C₁₀H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD24387418
CAS Number: 186312-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR