6-Bromo-2-chloro-3-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

60788-25G 60788-5G
76943-582EA 209 USD
76943-582 76943-584
6-Bromo-2-chloro-3-iodopyridine ≥97%
6-Bromo-2-chloro-3-iodopyridine
Formula: C₅H₂BrClIN
Storage Temperature: Ambient
MDL Number: MFCD11857690
CAS Number: 1138444-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR