Methyl 3-bromo-5-fluoro-2-methylbenzoate ≥95%
Supplier: AOBChem USA
|
Warning
|
60328-1G
60328-25G
60328-500MG
60328-5G
76943-572EA
159.14
USD
76943-572
76943-574
76943-576
76943-578
Methyl 3-bromo-5-fluoro-2-methylbenzoate ≥95%
Methyl 3-bromo-5-fluoro-2-methylbenzoate
Formula:
C₉H₈BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD19440363 CAS Number: 1187318-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...