Ethyl 2,6-difluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

60294-1G 60294-25G 60294-500MG 60294-5G
76943-558EA 124.13 USD
76943-558 76943-560 76943-562 76943-564
Ethyl 2,6-difluoro-4-methoxybenzoate ≥97%
Ethyl 2,6-difluoro-4-methoxybenzoate
Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD10566176
CAS Number: 1260849-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR