Methyl 3-formyl-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

59566-1G 59566-25G 59566-500MG 59566-5G
76943-532EA 352.22 USD
76943-532 76943-534 76943-536 76943-538
Methyl 3-formyl-5-methylbenzoate ≥97%
Methyl 3-formyl-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22544349
CAS Number: 1205514-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR