Methyl 3-chloro-4-hydroxy-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

59170-1G 59170-25G 59170-500MG 59170-5G
76943-516EA 406.33 USD
76943-516 76943-518 76943-520 76943-522
Methyl 3-chloro-4-hydroxy-5-methylbenzoate ≥97%
Methyl 3-chloro-4-hydroxy-5-methylbenzoate
Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
CAS Number: 883241-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR