6-Chloro-N-methoxy-N-methylpicolinamide ≥97%

Supplier: AOBChem USA

Warning

59054-1G 59054-25G 59054-500MG 59054-5G
76943-502EA 126.25 USD
76943-502 76943-504 76943-506 76943-508
6-Chloro-N-methoxy-N-methylpicolinamide ≥97%
6-Chloro-N-methoxy-N-methylpicolinamide
Formula: C₈H₉ClN₂O₂
MW: 200.622 g/mol
Storage Temperature: Ambient
CAS Number: 192437-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR