Ethyl 2-bromo-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

58508-1G 58508-25G 58508-5G
76943-480EA 76.39 USD
76943-480 76943-482 76943-484
Ethyl 2-bromo-5-methylbenzoate ≥97%
Ethyl 2-bromo-5-methylbenzoate
Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
CAS Number: 1210047-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR