6-Bromo-3-chloro-2-methylphenol ≥97% research grade

Supplier: AOBChem USA

Warning

58160-1G 58160-25G 58160-5G
76943-466EA 62.6 USD
76943-466 76943-468 76943-470
6-Bromo-3-chloro-2-methylphenol ≥97% research grade
6-Bromo-3-chloro-2-methylphenol
Formula: C₇H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD22414736
CAS Number: 55289-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR