Ethyl 6-bromo-2,3-difluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

57902-1G 57902-25G 57902-5G
76943-438EA 111.4 USD
76943-438 76943-440 76943-442
Ethyl 6-bromo-2,3-difluorobenzoate ≥97%
Ethyl 6-bromo-2,3-difluorobenzoate
Formula: C₉H₇BrF₂O₂
Storage Temperature: Refrigerator
CAS Number: 1807243-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR