6-Bromo-2-fluoro-3-methylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

57698-1G 57698-25G 57698-500MG 57698-5G
76943-410EA 391.48 USD
76943-410 76943-412 76943-414 76943-416
6-Bromo-2-fluoro-3-methylbenzonitrile ≥97%
6-Bromo-2-fluoro-3-methylbenzonitrile
Formula: C₈H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD26680100
CAS Number: 1501163-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR