Ethyl 3-hydroxy-3-(4-methoxyphenyl)propanoate ≥95%

Supplier: AOBChem USA

Warning

57588-25G 57588-5G
76943-398EA 595.17 USD
76943-398 76943-400
Ethyl 3-hydroxy-3-(4-methoxyphenyl)propanoate ≥95%
Ethyl 3-hydroxy-3-(4-methoxyphenyl)propanoate
Formula: C₁₂H₁₆O₄
MW: 224.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16756875
CAS Number: 70200-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR